Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MARK3 SIRNA Assays ›
Gene Symbol:
MARK3
Gene Name:
microtubule affinity regulating kinase 3
Gene Aliases:
CTAK1, KP78, PAR1A, Par-1a
Chromosome Location:
Chr.14: 103385364 - 103503831 on Build GRCh38
UniGene ID:
Hs.35828
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s230620
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MARK3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001128918.2
NP_001122390.2
13
2044
NM_001128919.2
NP_001122391.2
13
2044
NM_001128920.2
NP_001122392.2
13
1996
NM_001128921.2
NP_001122393.2
12
1807
NM_002376.6
NP_002367.5
13
2044
XM_005267641.4
XP_005267698.2
13
1423
XM_005267642.4
XP_005267699.2
13
1996
XM_005267643.4
XP_005267700.2
13
2044
XM_006720146.3
XP_006720209.1
13
1418
XM_011536769.2
XP_011535071.1
12
1157
XM_017021291.1
XP_016876780.1
13
1996
XM_017021292.1
XP_016876781.1
13
1996
XM_017021293.1
XP_016876782.1
14
1471
XM_017021294.1
XP_016876783.1
12
1793
XM_017021295.1
XP_016876784.1
12
1808
XM_017021296.1
XP_016876785.1
12
1825
XM_017021297.1
XP_016876786.1
12
1753
XM_017021298.1
XP_016876787.1
12
1801
XM_017021299.1
XP_016876788.1
12
1804
XM_017021300.1
XP_016876789.1
11
1278
XM_017021301.1
XP_016876790.1
11
1433
XM_017021302.1
XP_016876791.1
11
1278
XM_017021303.1
XP_016876792.1
11
1278
XM_017021304.1
XP_016876793.1
11
1278
XM_017021305.1
XP_016876794.1
11
1230
XM_017021306.1
XP_016876795.1
12
1973
XM_017021307.1
XP_016876796.1
11
1924
XM_017021308.1
XP_016876797.1
12
1947
XM_017021309.1
XP_016876798.1
11
1256
XM_017021310.1
XP_016876799.1
11
1256
XM_017021311.1
XP_016876800.1
10
1207
GenBank mRNA
AF170723.1
-
4
433
AF387637.1
-
13
1396
AF465413.1
-
13
1888
AK296643.1
-
6
633
AK307448.1
-
13
1423
BC024773.1
-
13
1867
5138958
BX161395.1
-
12
1737
BX248250.1
-
4
414
M80359.1
-
13
1501
U64205.1
-
13
1753
Back To Top
More Information
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
MAPK cascade
microtubule cytoskeleton organization
establishment of cell polarity
protein serine/threonine kinase activity
protein binding
ATP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0