Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other LOC613037 SIRNA Assays ›
          Gene Symbol:
          LOC613037 NPIPB11 NPIPB3 NPIPB5
          Gene Name:
          nuclear pore complex interacting protein member
          nuclear pore complex interacting protein family member B11
          nuclear pore complex interacting protein family member B3
          nuclear pore complex interacting protein family member B5
          Gene Aliases:
          -
          NPIP
          KIAA0220L, NPIP, NPIPB, NPIPB5, NPIPL3
          -
          Chromosome Location:
          Chr.16: 30222735 - 30251993 on Build GRCh38
          Chromosome Location:
          Chr.16: 29381318 - 29403802 on Build GRCh38
          Chromosome Location:
          Chr.16: 21402134 - 21425337 on Build GRCh38
          Chromosome Location:
          Chr.16: 22513523 - 22536521 on Build GRCh38
          UniGene ID:
          Hs.585733
          Hs.720286
          Hs.611072
          Hs.611072
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          siRNA ID s23070

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: LOC613037
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001321892.1 NP_001308821.1 4 397
          XM_017023514.1 XP_016879003.1 4 686
          XM_017023515.1 XP_016879004.1 5 705
          XM_017023516.1 XP_016879005.1 4 732
          XM_017023517.1 XP_016879006.1 4 463
          XM_017023518.1 XP_016879007.1 4 551
          XM_017023519.1 XP_016879008.1 4 608
          GenBank mRNA BQ433543.1 - 320
          DB231720.1 - 3 266
          Gene Symbol: NPIPB11
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001310137.1 NP_001297066.1 2 204
          GenBank mRNA BF592449.1 - 2 190
          KF175195.1 - 2 108
          KF175204.1 - 2 147
          Gene Symbol: NPIPB3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_130464.2 NP_569731.2 3 280
          GenBank mRNA AB209632.1 - 2 2847
          AK295170.1 - 3 279
          AK304162.1 - 1 62
          BC036263.1 - 3 280 3903371
          BC094882.1 - 2 207 3093162
          DC342292.1 - 2 384
          KF175178.1 - 2 105
          KF175179.1 - 2 147
          KF175180.1 - 2 147
          KF175181.1 - 2 291
          KF175182.1 - 2 147
          KF175183.1 - 1 39
          KF175184.1 - 1 30
          KF175187.1 - 2 147
          KF175188.1 - 2 108
          KF175190.1 - 2 147
          KF175191.1 - 2 108
          KF175192.1 - 2 108
          KF175193.1 - 2 111
          KF175194.1 - 2 111
          KF175196.1 - 2 108
          KF175197.1 - 2 108
          KF175198.1 - 2 204
          KF175199.1 - 2 204
          KF175200.1 - 2 204
          KF175201.1 - 2 147
          KF175202.1 - 2 108
          KF175203.1 - 2 204
          KF175205.1 - 2 204
          KF175206.1 - 2 204
          KF175215.1 - 4 466
          KF175216.1 - 3 481
          KF175218.1 - 2 109
          KF175223.1 - 2 204
          KF175224.1 - 2 147
          Gene Symbol: NPIPB5
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001135865.1 NP_001129337.1 2 483
          GenBank mRNA AK296338.1 - 2 324
          DC309028.1 - 2 483

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-2k58s:80/100.66.76.192:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0