Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NPIPB3 SIRNA Assays ›
          Gene Symbol:
          NPIPB3 LOC613037 NPIPB11 NPIPB4 NPIPB5
          Gene Name:
          nuclear pore complex interacting protein family member B3
          nuclear pore complex interacting protein member
          nuclear pore complex interacting protein family member B11
          nuclear pore complex interacting protein family member B4
          nuclear pore complex interacting protein family member B5
          Gene Aliases:
          KIAA0220L, NPIP, NPIPB, NPIPB5, NPIPL3
          -
          NPIP
          61E3.4
          -
          Chromosome Location:
          Chr.16: 21402134 - 21425337 on Build GRCh38
          Chromosome Location:
          Chr.16: 30222735 - 30251993 on Build GRCh38
          Chromosome Location:
          Chr.16: 29381318 - 29403802 on Build GRCh38
          Chromosome Location:
          Chr.16: 21834563 - 21859614 on Build GRCh38
          Chromosome Location:
          Chr.16: 22513523 - 22536521 on Build GRCh38
          UniGene ID:
          Hs.611072
          Hs.585733
          Hs.720286
          Hs.611072
          Hs.611072
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          siRNA ID s23071

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: NPIPB3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_130464.2 NP_569731.2 9 3086
          GenBank mRNA AB209632.1 - 11 4795
          AK128772.1 - 5 3831
          AK294452.1 - 5 2649
          AK295170.1 - 11 3211
          AK300277.1 - 2781
          AK303166.1 - 2, 3 2167
          AK303500.1 - 7 2706
          AK304162.1 - 6 3108
          AK316459.1 - 7 2641
          AY498718.1 - 1 13
          BC036263.1 - 9 1721 3903371
          BC061522.1 - 2 903 6190707
          BC073943.1 - 1 694
          BC094882.1 - 8 1888 3093162
          BM710413.1 - 1 247
          BQ950195.1 - 1 57
          Gene Symbol: LOC613037
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001321892.1 NP_001308821.1 9 3218
          XM_017023514.1 XP_016879003.1 9 3507
          XM_017023515.1 XP_016879004.1 10 3526
          XM_017023516.1 XP_016879005.1 9 3553
          XM_017023517.1 XP_016879006.1 9 3284
          XM_017023518.1 XP_016879007.1 9 3372
          XM_017023519.1 XP_016879008.1 9 3429
          XM_017023520.1 XP_016879009.1 4 4493
          XM_017023521.1 XP_016879010.1 5 2748
          XM_017023522.1 XP_016879011.1 2 3157
          Gene Symbol: NPIPB11
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001310137.1 NP_001297066.1 7 1405
          Gene Symbol: NPIPB4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001310148.1 NP_001297077.1 8 3502
          XM_017023244.1 XP_016878733.1 7 5513
          XM_017023245.1 XP_016878734.1 8 5350
          XM_017023246.1 XP_016878735.1 8 5271
          XM_017023247.1 XP_016878736.1 9 3258
          GenBank mRNA AK303607.1 - 2524
          Gene Symbol: NPIPB5
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001135865.1 NP_001129337.1 7 3289
          GenBank mRNA AK293635.1 - 1 2247
          AK296301.1 - 2903
          AK296338.1 - 7 3130
          AK303264.1 - 6 1732
          AK303572.1 - 2 2419
          AK316332.1 - 1 1537

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-st8h5:80/100.66.79.144:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0