Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TBC1D3H SIRNA Assays ›
          Gene Symbol:
          TBC1D3H LOC101060389 TBC1D3 TBC1D3B TBC1D3C TBC1D3F TBC1D3G TBC1D3I TBC1D3J TBC1D3K TBC1D3L
          Gene Name:
          TBC1 domain family member 3H
          TBC1 domain family member-like
          TBC1 domain family member 3
          TBC1 domain family member 3B
          TBC1 domain family member 3C
          TBC1 domain family member 3F
          TBC1 domain family member 3G
          TBC1 domain family member 3I
          TBC1 domain family member 3J
          TBC1 domain family member 3K
          TBC1 domain family member 3L
          Gene Aliases:
          -
          TBC1D3D
          PRC17, TBC1D3A, TBC1D3F
          PRC17, TBC1D3I, TBC1D3L
          TBC1D3D
          TBC1D3
          -
          -
          -
          -
          -
          Chromosome Location:
          Chr.17: 36377475 - 36388448 on Build GRCh38
          Chromosome Location:
          Chr.17: 38004033 - 38014908 on Build GRCh38
          Chromosome Location:
          Chr.17: 38181476 - 38192555 on Build GRCh38
          Chromosome Location:
          Chr.17: 36165681 - 36176624 on Build GRCh38
          Chromosome Location:
          Chr.17: 38057693 - 38068634 on Build GRCh38
          Chromosome Location:
          Chr.17: 36428612 - 36439523 on Build GRCh38
          Chromosome Location:
          Chr.17: 36322242 - 36334759 on Build GRCh38
          Chromosome Location:
          Chr.17: 36253639 - 36264601 on Build GRCh38
          Chromosome Location:
          Chr.17: 36450649 - 36458000 on Build GRCh38
          Chromosome Location:
          Chr.17: 37923805 - 37935371 on Build GRCh38
          Chromosome Location:
          Chr.17: 37978155 - 37990691 on Build GRCh38
          UniGene ID:
          Hs.105891
          Hs.454716
          Hs.454716
          Hs.105891
          Hs.735975
          Hs.617010
          Hs.454716
          Hs.454716
          -
          Hs.454716
          Hs.454716
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          siRNA ID s230757

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TBC1D3H
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001123392.3 NP_001116864.3 12 1002
          XM_006722055.1 XP_006722118.1 11 1642
          XM_006722056.1 XP_006722119.1 10 1576
          XM_011525172.1 XP_011523474.1 11 1825
          XM_011525173.2 XP_011523475.1 12 1196
          XM_011525174.1 XP_011523476.1 10 1759
          XM_011525175.1 XP_011523477.1 11 1825
          XM_011525176.1 XP_011523478.1 11 1825
          GenBank mRNA BM844606.1 - 3 200
          BX281608.1 - 6 448
          Gene Symbol: LOC101060389
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001291465.1 NP_001278394.1 12 988
          XM_006722248.2 XP_006722311.1 11 2494
          XM_006722249.2 XP_006722312.1 10 2428
          XM_006722250.2 XP_006722313.1 11 2494
          XM_006722252.3 XP_006722315.1 10 2245
          XM_006722253.2 XP_006722316.1 11 2494
          XM_011524176.2 XP_011522478.1 12 1101
          Gene Symbol: TBC1D3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001123391.3 NP_001116863.3 12 1000
          GenBank mRNA AK302064.1 - 13 1226
          AK303259.1 - 11 1020
          AX775941.1 - 14 1249
          AX775943.1 - 14 1249
          Gene Symbol: TBC1D3B
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001001417.6 NP_001001417.6 12 1034
          XM_005257980.4 XP_005258037.2 12 1164
          GenBank mRNA AF540953.1 - 11 888
          AK292257.1 - 12 1005
          Gene Symbol: TBC1D3C
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001001418.6 NP_001001418.5 12 1028
          XM_006721904.2 XP_006721967.1 11 2494
          XM_006721905.2 XP_006721968.1 10 2428
          XM_006721907.3 XP_006721970.1 10 2245
          XM_006721908.2 XP_006721971.1 11 2494
          XM_011524814.2 XP_011523116.1 12 1101
          XM_017024646.1 XP_016880135.1 11 2494
          Gene Symbol: TBC1D3F
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_032258.4 NP_115634.3 12 1002
          GenBank mRNA AK225985.1 - 12 993
          AK292344.1 - 12 1045
          BC075809.1 - 12 1034 5170064
          Gene Symbol: TBC1D3G
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001291462.1 NP_001278391.1 12 988
          XM_005276914.3 XP_005276971.1 12 2813
          GenBank mRNA AK292308.1 - 13 1038
          AK301823.1 - 10 800
          BC071680.1 - 12 988 5165385
          Gene Symbol: TBC1D3I
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001291463.1 NP_001278392.1 12 988
          XM_006722223.3 XP_006722286.1 12 1259
          XM_006722224.3 XP_006722287.1 11 2504
          XM_006722226.3 XP_006722289.1 10 2438
          XM_011524183.2 XP_011522485.1 10 2621
          XM_011524184.2 XP_011522486.1 11 2687
          XM_011524185.2 XP_011522487.1 11 2687
          Gene Symbol: TBC1D3J
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_017025494.1 XP_016880983.1 10 804
          Gene Symbol: TBC1D3K
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001291464.1 NP_001278393.1 12 988
          XM_006722235.2 XP_006722298.1 11 3266
          XM_006722236.1 XP_006722299.1 12 1838
          XM_006722237.2 XP_006722300.1 11 3083
          XM_006722239.2 XP_006722302.1 10 3017
          XM_011524172.1 XP_011522474.1 10 3200
          XM_011524173.1 XP_011522475.1 11 3266
          XM_011524174.1 XP_011522476.1 11 3266
          Gene Symbol: TBC1D3L
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001291459.1 NP_001278388.1 12 1002
          XM_006722242.3 XP_006722305.1 11 4242
          XM_006722243.2 XP_006722306.1 10 2428
          XM_006722244.2 XP_006722307.1 11 2494
          XM_006722246.3 XP_006722309.1 10 1576
          XM_006722247.2 XP_006722310.1 11 2494
          XM_011524175.2 XP_011522477.1 12 1101
          GenBank mRNA AK095385.1 - 10 1576
          AK122833.1 - 8 2313
          AX775949.1 - 10 1576
          AX775951.1 - 10 1576

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          GTPase-activating protein

          Gene Ontology Categories:

          Function(s) Process(es)

          intracellular protein transport
          regulation of vesicle fusion
          activation of GTPase activity
          positive regulation of GTPase activity
          regulation of cilium assembly
          GTPase activator activity
          Rab GTPase binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0