Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MICAL3 SIRNA Assays ›
          Gene Symbol:
          MICAL3
          Gene Name:
          microtubule associated monooxygenase, calponin and LIM domain containing 3
          Gene Aliases:
          MICAL-3
          Chromosome Location:
          Chr.22: 17787649 - 18024559 on Build GRCh38
          UniGene ID:
          Hs.528024
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s230763

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: MICAL3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001122731.2 NP_001116203.1 3, 4 658
          NM_001136004.3 NP_001129476.1 3, 4 658
          NM_015241.2 NP_056056.2 4, 5 937
          XM_005261262.3 XP_005261319.1 4, 5 767
          XM_011546137.2 XP_011544439.1 4, 5 767
          XM_011546139.2 XP_011544441.1 4, 5 767
          XM_011546142.2 XP_011544444.1 4, 5 767
          XM_011546146.2 XP_011544448.1 4, 5 767
          XM_017028865.1 XP_016884354.1 4, 5 1285
          XM_017028866.1 XP_016884355.1 4, 5 1091
          XM_017028867.1 XP_016884356.1 4, 5 2208
          XM_017028868.1 XP_016884357.1 4, 5 937
          XM_017028869.1 XP_016884358.1 4, 5 682
          XM_017028870.1 XP_016884359.1 4, 5 683
          XM_017028871.1 XP_016884360.1 4, 5 767
          XM_017028872.1 XP_016884361.1 4, 5 767
          XM_017028873.1 XP_016884362.1 4, 5 767
          XM_017028874.1 XP_016884363.1 4, 5 767
          XM_017028875.1 XP_016884364.1 4, 5 767
          XM_017028876.1 XP_016884365.1 4, 5 767
          XM_017028877.1 XP_016884366.1 4, 5 767
          XM_017028878.1 XP_016884367.1 4, 5 767
          XM_017028879.1 XP_016884368.1 4, 5 767
          XM_017028880.1 XP_016884369.1 4, 5 1090
          XM_017028881.1 XP_016884370.1 4, 5 937
          XM_017028882.1 XP_016884371.1 4, 5 767
          XM_017028883.1 XP_016884372.1 2, 3 322
          XM_017028884.1 XP_016884373.1 4, 5 1108
          XM_017028887.1 XP_016884376.1 4, 5 767
          GenBank mRNA AB037785.1 - 1, 2 89
          AK127487.1 - 4, 5 767
          AK295978.1 - 4, 5 909
          BC157876.1 - 3, 4 646 8862700
          BC171887.1 - 3, 4 646 9054541
          BX647382.1 - 3, 4 607
          CR456364.1 - 3, 4 599

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Mouse: NM_153396, NM_001270475, AK132669, AK122497, AF536756, and others.

          Gene Ontology Categories:

          Function(s) Process(es)

          exocytosis
          cytoskeleton organization
          actin filament depolymerization
          oxidation-reduction process
          actin binding
          zinc ion binding
          oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen, NAD(P)H as one donor, and incorporation of one atom of oxygen
          Rab GTPase binding
          FAD binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-qvstf:80/100.66.73.196:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0