Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NSUN5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168347.2 | NP_001161819.1 | 6, 7 | 840 | ||
NM_001168348.2 | NP_001161820.1 | 6, 7 | 726 | |||
NM_018044.4 | NP_060514.1 | 6, 7 | 840 | |||
NM_148956.3 | NP_683759.1 | 6, 7 | 840 | |||
GenBank mRNA | AF412028.1 | - | 6, 7 | 779 | ||
AF420249.1 | - | 6, 7 | 771 | |||
AK001129.1 | - | 6, 7 | 779 | |||
AK098619.1 | - | 6, 7 | 799 | |||
AK126375.1 | - | 6, 7 | 663 | |||
AK298221.1 | - | 6, 7 | 789 | |||
AK315361.1 | - | 6, 7 | 781 | |||
BC008084.2 | - | 6, 7 | 780 | 2823604 | ||
CN418523.1 | - | 2, 3 | 164 |
Gene Symbol: NSUN5P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033322.3 | 7, 8 | 938 | |||
NR_104013.1 | 6, 7 | 771 | ||||
GenBank mRNA | AF416610.1 | - | 6, 7 | 802 | ||
AK094312.1 | - | 2, 3 | 1714 | |||
AK126486.1 | - | 4, 5 | 3688 | |||
AK128374.1 | - | 2, 3 | 1926 | |||
BG741008.1 | - | 4, 5 | 480 |
Gene Symbol: NSUN5P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033323.3 | 7, 8 | 939 | |||
GenBank mRNA | AF416611.1 | - | 7, 8 | 896 | ||
AF420250.1 | - | 7, 8 | 896 | |||
AK021688.1 | - | 7, 8 | 896 | |||
AK292107.1 | - | 3, 4 | 372 | |||
AK303622.1 | - | 2, 3 | 718 | |||
AK315927.1 | - | 2, 3 | 552 | |||
AL833016.1 | - | 6, 7 | 1582 | |||
BC007488.2 | - | 7, 8 | 993 | |||
BC082753.1 | - | 3, 4 | 352 | 4542435 | ||
BC106049.1 | - | 3, 4 | 420 |