Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NSUN5P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033323.3 | 3 | 275 | |||
GenBank mRNA | AF416611.1 | - | 3 | 232 | ||
AF420250.1 | - | 3 | 232 | |||
AK021688.1 | - | 3 | 232 | |||
AL833016.1 | - | 2 | 918 | |||
BC007488.2 | - | 3 | 329 | |||
DA017627.1 | - | 3 | 383 |
Gene Symbol: NSUN5P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033322.3 | 3 | 275 | |||
GenBank mRNA | AF416610.1 | - | 2 | 139 | ||
AK126486.1 | - | 1 | 1073 | |||
DB270164.1 | - | 3 | 258 |