siRNA Details
Gene Symbol: SNRPN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003097.4 | NP_003088.1 | 5 | 598 | ||
NM_022805.3 | NP_073716.1 | 7 | 793 | |||
NM_022806.3 | NP_073717.1 | 7 | 803 | |||
NM_022807.3 | NP_073718.1 | 8 | 938 | |||
NM_022808.3 | NP_073719.1 | 7 | 830 | |||
GenBank mRNA | AF400432.1 | - | 2 | 53 | ||
AK123119.1 | - | 3 | 1089 | |||
AK222633.1 | - | 7 | 753 | |||
BC000611.2 | - | 2 | 224 | 3347412 | ||
BC003180.1 | - | 5 | 504 | 2958068 | ||
BC018809.2 | - | 5 | 493 | |||
BC024777.1 | - | 6 | 626 | 3907606 | ||
BC025178.1 | - | 5 | 470 | 4158469 | ||
BC035280.1 | - | 9 | 972 | |||
CR450350.1 | - | 1 | 38 | |||
J04615.1 | - | 5 | 503 | |||
KJ534946.1 | - | 1 | 38 | |||
L80004.1 | - | 7 | 782 | |||
L80005.1 | - | 8 | 928 | |||
U41303.1 | - | 5 | 503 | |||
X15892.1 | - | 3 | 380 |
Gene Symbol: SNURF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005678.4 | NP_005669.2 | 5 | 598 | ||
GenBank mRNA | AK098563.1 | - | 5 | 502 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
Molecular Function -




