Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZACN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_180990.3 | NP_851321.2 | 8 | 1000 | ||
GenBank mRNA | AB223030.1 | - | 8 | 914 | ||
AF512521.1 | - | 8 | 954 | |||
AK122638.1 | - | 8 | 1694 | |||
BC110596.1 | - | 6 | 956 | |||
BC110597.1 | - | 8 | 954 | 40023955 |