Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DEFB103B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018661.4 | NP_061131.1 | 1 | 86 | ||
GenBank mRNA | AF301470.1 | - | 1 | 86 |
Gene Symbol: DEFB103A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081551.3 | NP_001075020.1 | 1 | 86 |