Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007099.2 | NP_001007100.1 | 5 | 632 | ||
NM_001007100.2 | NP_001007101.1 | 5 | 623 | |||
NM_001007250.2 | NP_001007251.1 | 4 | 502 | |||
NM_001193599.1 | NP_001180528.1 | 15 | 1857 | |||
NM_001193600.1 | NP_001180529.1 | 15 | 1797 | |||
NM_001193617.1 | NP_001180546.1 | 15 | 1871 | |||
NM_002979.4 | NP_002970.2 | 16 | 1929 | |||
XM_005271103.4 | XP_005271160.1 | 15 | 1742 | |||
XM_017002046.1 | XP_016857535.1 | 6 | 1197 | |||
GenBank mRNA | AB208789.1 | - | 17 | 4624 | ||
AK294631.1 | - | 15 | 1800 | |||
AK295214.1 | - | 15 | 1814 | |||
AK308105.1 | - | 15 | 1739 | |||
BC005911.1 | - | 5 | 622 | |||
BX537618.1 | - | 3 | 3138 | |||
BX537619.1 | - | 3 | 3138 | |||
CA867670.1 | - | 4 | 459 | |||
S52450.1 | - | 5 | 632 |