Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_170723.1 | NP_733841.1 | 1 | 2050 | ||
GenBank mRNA | AK292375.1 | - | 1 | 2072 | ||
BC033041.1 | - | 1 | 2055 |
Gene Symbol: CDY1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003894.1 | NP_001003894.1 | 1 | 2050 |