siRNA Details
Gene Symbol: KCNK16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135105.1 | NP_001128577.1 | 2 | 330 | ||
NM_001135106.1 | NP_001128578.1 | 2 | 330 | |||
NM_001135107.1 | NP_001128579.1 | 2 | 330 | |||
NM_032115.3 | NP_115491.1 | 2 | 330 | |||
XM_011514935.2 | XP_011513237.1 | 3 | 386 | |||
XM_017011346.1 | XP_016866835.1 | 2 | 424 | |||
GenBank mRNA | AY253145.1 | - | 2 | 316 | ||
AY253146.1 | - | 2 | 316 | |||
BC111860.1 | - | 2 | 316 | 40016308 | ||
CA843225.1 | - | 2 | 329 | |||
EU978943.1 | - | 2 | 364 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
