Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRTAP6-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181602.1 | NP_853633.1 | 1 | 53 | ||
GenBank mRNA | AB096954.1 | - | 1 | 79 |