Search
Search

FAM175B| Gene Symbol: FAM175B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032182.3 | NP_115558.3 | 9 | 1095 | ||
| XM_011539554.1 | XP_011537856.1 | 8 | 999 | |||
| XM_017015986.1 | XP_016871475.1 | 7 | 973 | |||
| GenBank mRNA | AK025991.1 | - | 2 | 377 | ||
| AK296677.1 | - | 9 | 1067 | |||
| BC008999.1 | - | 9 | 1057 | 2900632 | ||
| D63877.1 | - | 9 | 1064 | |||