Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other HIST1H2BK SIRNA Assays ›
          Gene Symbol:
          Hist1h2bk Hist1h2bc Hist1h2be Hist1h2bf Hist1h2bg Hist1h2bh Hist1h2bj Hist1h2bl Hist1h2bn Hist1h2bq Hist1h2br
          Gene Name:
          histone cluster 1, H2bk
          histone cluster 1, H2bc
          histone cluster 1, H2be
          histone cluster 1, H2bf
          histone cluster 1, H2bg
          histone cluster 1, H2bh
          histone cluster 1, H2bj
          histone cluster 1, H2bl
          histone cluster 1, H2bn
          histone cluster 1, H2bq
          histone cluster 1 H2br
          Gene Aliases:
          -
          2610022J01Rik, H2bfs, R74621
          -
          -
          -
          H2b-221
          -
          H2b-291a
          -
          Gm13646, OTTMUSG00000013203
          Gm11277, OTTMUSG00000000449
          Chromosome Location:
          Chr.13: 22035821 - 22036320 on Build GRCm38
          Chromosome Location:
          Chr.13: 23684199 - 23692480 on Build GRCm38
          Chromosome Location:
          Chr.13: 23583670 - 23621124 on Build GRCm38
          Chromosome Location:
          Chr.13: 23573760 - 23574190 on Build GRCm38
          Chromosome Location:
          Chr.13: 23571400 - 23571863 on Build GRCm38
          Chromosome Location:
          Chr.13: 23542923 - 23543444 on Build GRCm38
          Chromosome Location:
          Chr.13: 22043230 - 22043658 on Build GRCm38
          Chromosome Location:
          Chr.13: 21715713 - 21716143 on Build GRCm38
          Chromosome Location:
          Chr.13: 21754123 - 21754553 on Build GRCm38
          Chromosome Location:
          Chr.13: 21806412 - 21810199 on Build GRCm38
          Chromosome Location:
          Chr.13: 21833743 - 21837530 on Build GRCm38
          UniGene ID:
          Mm.358955
          Mm.261676
          Mm.440391
          -
          Mm.261673
          Mm.352273
          Mm.371766
          -
          -
          Mm.371766
          Mm.471247
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          siRNA ID s232317

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 404.00
          Your Price
          Online offer: 391.65
          391.65
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Hist1h2bk
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_175665.2 NP_783596.1 1 142
          GenBank mRNA BC147143.1 - 1 142 9056184
          BC147144.1 - 1 142 9056197
          Gene Symbol: Hist1h2bc
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001290380.1 NP_001277309.1 1 109
          NM_023422.3 NP_075911.2 1 109
          GenBank mRNA AK005191.1 - 1 104
          AK005407.1 - 1 104
          AK011516.1 - 1 111
          BC019673.1 - 1 92 3993954
          BC082774.1 - 1 92
          Gene Symbol: Hist1h2be
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001177653.1 NP_001171124.1 2 269
          NM_001290530.1 NP_001277459.1 1 90
          NM_178194.4 NP_835501.1 2 295
          GenBank mRNA AK030546.1 - 2 297
          AK049948.1 - 2 270
          BC069889.1 - 1 83 4217814
          Gene Symbol: Hist1h2bf
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_178195.2 NP_835502.1 1 76
          Gene Symbol: Hist1h2bg
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_178196.4 NP_835503.1 1 109
          GenBank mRNA BC060304.1 - 1 136 4217587
          Gene Symbol: Hist1h2bh
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_178197.2 NP_835504.1 1 168
          GenBank mRNA AK158416.1 - 117
          BC092138.1 - 83 30613720
          BC132584.1 - 1 168 40130861
          BC132586.1 - 1 168 40130863
          Gene Symbol: Hist1h2bj
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_178198.2 NP_835505.1 1 76
          GenBank mRNA BC115792.1 - 68
          Gene Symbol: Hist1h2bl
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_178199.2 NP_835506.1 1 76
          Gene Symbol: Hist1h2bn
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_178201.2 NP_835508.1 1 76
          Gene Symbol: Hist1h2bq
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001097979.2 NP_001091448.2 1 104
          Gene Symbol: Hist1h2br
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001110555.1 NP_001104025.1 1 104
          GenBank mRNA AK036389.1 - 1 105
          BC011440.1 - 1 89 3989862

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Human: NM_003520, BC101411, AK312228, BC101413, BC101412.
          This siRNA also targets the following transcripts in Rat: XM_008771682.

          Panther Classification:

          Molecular Function -

          histone

          Gene Ontology Categories:

          Function(s) Process(es)

          innate immune response in mucosa
          nucleosome assembly
          antibacterial humoral response
          defense response to Gram-positive bacterium
          biological_process
          DNA binding
          protein heterodimerization activity
          molecular_function

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0