Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Western Blot Products
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Food and Beverage
Lab Solutions
Pharma and Biopharma
Real-Time PCR
Semiconductor Analysis
Clinical and Diagnostics
Digital Solutions
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
See all services
Help and Support
Order Help
Digital Solutions
Product Support
Technical Information
Training and Education
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PHLDB1 SIRNA Assays ›
Gene Symbol:
PHLDB1
Gene Name:
pleckstrin homology like domain family B member 1
Gene Aliases:
LL5A
Chromosome Location:
Chr.11: 118606296 - 118658038 on Build GRCh38
UniGene ID:
Hs.504062
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s23233
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PHLDB1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001144758.2
NP_001138230.1
6
1877
NM_001144759.2
NP_001138231.1
6
1877
NM_015157.3
NP_055972.1
7
2100
XM_005271468.3
XP_005271525.1
6
2646
XM_005271469.3
XP_005271526.1
6
2646
XM_005271470.3
XP_005271527.1
6
2646
XM_005271471.3
XP_005271528.1
6
2646
XM_005271472.3
XP_005271529.1
6
2646
XM_005271473.3
XP_005271530.1
6
2646
XM_005271474.4
XP_005271531.1
6
2646
XM_005271475.3
XP_005271532.1
6
2646
XM_005277680.3
XP_005277737.1
6
2643
XM_006718796.3
XP_006718859.1
6
1832
XM_006718797.3
XP_006718860.1
6
2646
XM_006718798.3
XP_006718861.1
6
2646
XM_006718799.3
XP_006718862.1
6
2646
XM_006718800.3
XP_006718863.1
6
2646
XM_006718801.3
XP_006718864.1
6
2646
XM_006718802.3
XP_006718865.1
6
2646
XM_006718803.3
XP_006718866.1
6
2646
XM_011542703.2
XP_011541005.1
6
2646
XM_011542704.2
XP_011541006.1
6
2646
XM_011542705.2
XP_011541007.1
6
2646
XM_011542706.2
XP_011541008.1
6
2646
XM_011542707.2
XP_011541009.1
6
2646
XM_011542708.2
XP_011541010.1
6
1835
XM_011542709.2
XP_011541011.1
6
2145
XM_011542710.2
XP_011541012.1
6
2646
XM_011542711.2
XP_011541013.1
3
1552
XM_011542713.2
XP_011541015.1
3
1546
XM_011542714.2
XP_011541016.1
6
2646
XM_017017407.1
XP_016872896.1
6
2643
XM_017017408.1
XP_016872897.1
6
2646
XM_017017409.1
XP_016872898.1
6
2646
XM_017017410.1
XP_016872899.1
6
2646
XM_017017411.1
XP_016872900.1
6
2646
GenBank mRNA
AB014538.2
-
6
1854
AB094090.1
-
7
2100
AB177859.1
-
1
981
AK074070.2
-
6
1826
AL832373.1
-
6
1834
BC098586.1
-
6
1854
30342198
CR749783.1
-
7
2013
Back To Top
More Information
Gene Ontology Categories:
Function(s)
regulation of gastrulation
regulation of epithelial to mesenchymal transition
regulation of microtubule cytoskeleton organization
positive regulation of basement membrane assembly involved in embryonic body morphogenesis
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0