Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYFIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033028.1 | NP_001028200.1 | 4 | 1139 | ||
NM_001287810.2 | NP_001274739.1 | 20 | 2410 | |||
NM_001324119.1 | NP_001311048.1 | 19 | 2239 | |||
NM_001324120.1 | NP_001311049.1 | 19 | 2418 | |||
NM_001324122.1 | NP_001311051.1 | 20 | 2439 | |||
NM_001324123.1 | NP_001311052.1 | 19 | 2318 | |||
NM_001324124.1 | NP_001311053.1 | 18 | 2201 | |||
NM_001324125.1 | NP_001311054.1 | 16 | 2000 | |||
NM_001324126.1 | NP_001311055.1 | 18 | 2189 | |||
NM_014608.4 | NP_055423.1 | 19 | 2291 | |||
XM_011543873.2 | XP_011542175.1 | 19 | 2640 | |||
XM_011543874.1 | XP_011542176.1 | 19 | 2673 | |||
XM_011543876.2 | XP_011542178.2 | 19 | 2241 | |||
XM_017022023.1 | XP_016877512.1 | 19 | 2646 | |||
XM_017022024.1 | XP_016877513.1 | 19 | 2654 | |||
GenBank mRNA | AK125936.1 | - | 18 | 2344 | ||
AK127094.1 | - | 4 | 1139 | |||
AK291604.1 | - | 19 | 2218 | |||
AL357618.1 | - | 3 | 283 | |||
AY763577.1 | - | 19 | 2255 | |||
AY763578.1 | - | 20 | 2321 | |||
AY763579.1 | - | 4 | 1139 | |||
BC001306.2 | - | 2 | 107 | |||
BC005097.1 | - | 19 | 2182 | 3163591 | ||
BC063869.1 | - | 4 | 933 | |||
D38549.1 | - | 19 | 2185 | |||
KJ534819.1 | - | 18 | 2130 |