Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other UCN SIRNA Assays ›
          Gene Symbol:
          Ucn
          Gene Name:
          urocortin
          Gene Aliases:
          Mpv17, Ucn1
          Chromosome Location:
          Chr.5: 31137989 - 31138895 on Build GRCm38
          UniGene ID:
          Mm.377116
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID s232507

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Ucn
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_021290.2 NP_067265.1 2 450
          GenBank mRNA BC116948.1 - 2 384 40126267
          BC116950.1 - 2 384 40126269
          BC144891.1 - 2 379 9053439

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_019150, U33935.

          Panther Classification:

          Molecular Function -

          peptide hormone

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of protein phosphorylation
          startle response
          response to oxidative stress
          neuropeptide signaling pathway
          sensory perception of sound
          learning or memory
          feeding behavior
          associative learning
          aerobic respiration
          positive regulation of gene expression
          negative regulation of gene expression
          response to auditory stimulus
          pancreatic juice secretion
          positive regulation of cell growth
          positive regulation of cAMP biosynthetic process
          negative regulation of histone deacetylation
          neuron projection development
          negative regulation of appetite
          positive regulation of interleukin-6 production
          positive regulation of collagen biosynthetic process
          positive regulation of peptidyl-serine phosphorylation
          activation of protein kinase A activity
          social behavior
          gastric emptying
          response to immobilization stress
          response to starvation
          drinking behavior
          negative regulation of apoptotic process
          negative regulation of catalytic activity
          positive regulation of vascular permeability
          positive regulation of translation
          positive regulation of DNA replication
          negative regulation of blood pressure
          negative regulation of cell size
          positive regulation of vasodilation
          positive regulation of transcription from RNA polymerase II promoter
          negative regulation of hormone secretion
          positive regulation of corticotropin secretion
          regulation of synaptic transmission, glutamatergic
          prepulse inhibition
          positive regulation of cardiac muscle contraction
          negative regulation of gastric acid secretion
          negative regulation of necrotic cell death
          negative regulation of cell death
          positive regulation of calcium ion import
          negative regulation of neuron death
          negative regulation of feeding behavior
          positive regulation of behavioral fear response
          G-protein coupled receptor binding
          hormone activity
          histone deacetylase inhibitor activity
          corticotropin-releasing hormone receptor 1 binding
          corticotropin-releasing hormone receptor 2 binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0