Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MDN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014611.2 | NP_055426.1 | 97 | 16233 | ||
XM_005248700.3 | XP_005248757.1 | 58 | 10165 | |||
XM_006715405.3 | XP_006715468.1 | 61 | 11711 | |||
XM_011535635.2 | XP_011533937.1 | 97 | 16038 | |||
GenBank mRNA | AB002299.2 | - | 51 | 8889 | ||
AF503925.1 | - | 97 | 16162 | |||
BC020432.1 | - | 3 | 216 | |||
BC036888.1 | - | 2 | 73 | |||
CR627399.1 | - | 43 | 7688 |