Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other HIST1H2BK SIRNA Assays ›
          Gene Symbol:
          Hist1h2bk Hist1h2bb Hist1h2bc Hist1h2be Hist1h2bf Hist1h2bg Hist1h2bh Hist1h2bj Hist1h2bl Hist1h2bm Hist1h2bn Hist1h2bp Hist1h2bq Hist1h2br
          Gene Name:
          histone cluster 1, H2bk
          histone cluster 1, H2bb
          histone cluster 1, H2bc
          histone cluster 1, H2be
          histone cluster 1, H2bf
          histone cluster 1, H2bg
          histone cluster 1, H2bh
          histone cluster 1, H2bj
          histone cluster 1, H2bl
          histone cluster 1, H2bm
          histone cluster 1, H2bn
          histone cluster 1, H2bp
          histone cluster 1, H2bq
          histone cluster 1 H2br
          Gene Aliases:
          -
          H2b-143
          2610022J01Rik, H2bfs, R74621
          -
          -
          -
          H2b-221
          -
          H2b-291a
          H2b-291b
          -
          -
          Gm13646, OTTMUSG00000013203
          Gm11277, OTTMUSG00000000449
          Chromosome Location:
          Chr.13: 22035821 - 22036320 on Build GRCm38
          Chromosome Location:
          Chr.13: 23746734 - 23747223 on Build GRCm38
          Chromosome Location:
          Chr.13: 23684199 - 23692480 on Build GRCm38
          Chromosome Location:
          Chr.13: 23583670 - 23621124 on Build GRCm38
          Chromosome Location:
          Chr.13: 23573760 - 23574190 on Build GRCm38
          Chromosome Location:
          Chr.13: 23571400 - 23571863 on Build GRCm38
          Chromosome Location:
          Chr.13: 23542923 - 23543444 on Build GRCm38
          Chromosome Location:
          Chr.13: 22043230 - 22043658 on Build GRCm38
          Chromosome Location:
          Chr.13: 21715713 - 21716143 on Build GRCm38
          Chromosome Location:
          Chr.13: 21722044 - 21722526 on Build GRCm38
          Chromosome Location:
          Chr.13: 21754123 - 21754553 on Build GRCm38
          Chromosome Location:
          Chr.13: 21787480 - 21801561 on Build GRCm38
          Chromosome Location:
          Chr.13: 21806412 - 21810199 on Build GRCm38
          Chromosome Location:
          Chr.13: 21833743 - 21837530 on Build GRCm38
          UniGene ID:
          Mm.358955
          Mm.390567
          Mm.261676
          Mm.440391
          -
          Mm.261673
          Mm.352273
          Mm.371766
          -
          Mm.277425
          -
          Mm.264645
          Mm.371766
          Mm.471247
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          siRNA ID s232562

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Hist1h2bk
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_175665.2 NP_783596.1 1 190
          GenBank mRNA BC147143.1 - 1 190 9056184
          BC147144.1 - 1 190 9056197
          Gene Symbol: Hist1h2bb
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA BC116853.1 - 1 185 40126172
          BC116857.1 - 1 185 40126176
          Gene Symbol: Hist1h2bc
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001290380.1 NP_001277309.1 1 157
          NM_023422.3 NP_075911.2 1 157
          GenBank mRNA AK005191.1 - 1 153
          AK005407.1 - 1 152
          AK011516.1 - 1 159
          BC019673.1 - 1 140 3993954
          BC082774.1 - 1 140
          Gene Symbol: Hist1h2be
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001177653.1 NP_001171124.1 2 317
          NM_001290530.1 NP_001277459.1 1 138
          NM_178194.4 NP_835501.1 2 343
          GenBank mRNA AK030546.1 - 2 345
          AK049948.1 - 2 318
          BC069889.1 - 1 131 4217814
          Gene Symbol: Hist1h2bf
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_178195.2 NP_835502.1 1 124
          Gene Symbol: Hist1h2bg
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_178196.4 NP_835503.1 1 157
          GenBank mRNA BC060304.1 - 1 184 4217587
          Gene Symbol: Hist1h2bh
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_178197.2 NP_835504.1 1 216
          GenBank mRNA AK158416.1 - 165
          BC092138.1 - 131 30613720
          BC132584.1 - 1 216 40130861
          BC132586.1 - 1 216 40130863
          Gene Symbol: Hist1h2bj
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA BC115792.1 - 116
          Gene Symbol: Hist1h2bl
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_178199.2 NP_835506.1 1 124
          Gene Symbol: Hist1h2bm
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_178200.2 NP_835507.1 1 178
          GenBank mRNA BC139381.1 - 1 178 8862403
          BC139382.1 - 1 178 8862404
          Gene Symbol: Hist1h2bn
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_178201.2 NP_835508.1 1 124
          Gene Symbol: Hist1h2bp
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001290466.1 NP_001277395.1 1 151
          NM_178202.2 NP_835509.2 1 151
          GenBank mRNA BC061044.1 - 1 151 6742370
          M25487.1 - 1 112
          Gene Symbol: Hist1h2bq
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001097979.2 NP_001091448.2 1 152
          Gene Symbol: Hist1h2br
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001110555.1 NP_001104025.1 1 152
          GenBank mRNA AK036389.1 - 1 153
          BC011440.1 - 1 137 3989862

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001109400, NM_022647, NM_001106114, NM_001108414, XM_225384, and others.

          Panther Classification:

          Molecular Function -

          histone

          Gene Ontology Categories:

          Function(s) Process(es)

          innate immune response in mucosa
          nucleosome assembly
          antibacterial humoral response
          defense response to Gram-positive bacterium
          biological_process
          DNA binding
          protein heterodimerization activity
          molecular_function

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0