Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rhox2h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100465.1 | NP_001093935.1 | 2 | 415 |
Gene Symbol: Rhox2f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085356.1 | NP_001078825.1 | 2 | 415 |