Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Jmjd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114637.1 | NP_001108109.1 | 3 | 408 | ||
GenBank mRNA | BC016255.1 | - | 3 | 385 |
Gene Symbol: Gm28042 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB195274.1 | - | 3 | 398 | ||
AB195275.1 | - | 3 | 398 |