Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhrs7c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013013.2 | NP_001013031.2 | 4 | 606 | ||
GenBank mRNA | AI391290.1 | - | 1 | 95 | ||
BC039565.1 | - | 4 | 497 | |||
CJ048402.1 | - | 4 | 602 |