Search
Search

Dlx6| Gene Symbol: Dlx6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010057.2 | NP_034187.1 | 2 | 512 | ||
| GenBank mRNA | AB221684.1 | - | 2 | 100 | ||
| AF022078.1 | - | 2 | 49 | |||
| BC114342.1 | - | 2 | 96 | |||
| EF535989.1 | - | 2 | 512 | |||
| EF535990.1 | - | 3 | 367 | |||