Search
Search

Eprs| Gene Symbol: Eprs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029735.1 | NP_084011.1 | 5 | 514 | ||
| GenBank mRNA | AK010734.1 | - | 5 | 478 | ||
| AK088974.1 | - | 5 | 543 | |||
| AK141072.1 | - | 5 | 566 | |||
| AK147995.1 | - | 5 | 478 | |||
| AK148463.1 | - | 5 | 475 | |||
| AK160986.1 | - | 5 | 535 | |||
| AK164073.1 | - | 4 | 2425 | |||
| AK165637.1 | - | 4 | 307 | |||
| AK201764.1 | - | 3 | 221 | |||
| BC040802.1 | - | 5 | 512 | |||
| BC058337.1 | - | 5 | 467 | |||
| BC094679.1 | - | 4 | 308 | |||
| BC141049.1 | - | 5 | 433 | 9055661 | ||
| CB520692.1 | - | 4 | 326 | |||
| CF537682.1 | - | 5 | 514 | |||