Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Defa-rs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007844.2 | NP_031870.2 | 2 | 355 | ||
GenBank mRNA | AK008584.1 | - | 2 | 356 | ||
BC117042.1 | - | 2 | 330 | 40126361 | ||
BC117044.1 | - | 2 | 330 | 40126363 | ||
BC145556.1 | - | 2 | 360 | 9054120 | ||
BC171966.1 | - | 2 | 360 | 9054252 | ||
EU760893.1 | - | 1 | 151 | |||
EU760894.1 | - | 1 | 151 | |||
M33226.1 | - | 2 | 335 |
Gene Symbol: Gm14851 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177482.1 | NP_001170953.1 | 2 | 310 | ||
GenBank mRNA | AV063918.1 | - | 1 | 145 | ||
AV064248.1 | - | 1 | 146 | |||
AY761183.1 | - | 2 | 310 |