Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other IFNZ SIRNA Assays ›
          Gene Symbol:
          Ifnz Gm13272 Gm13276 Gm13277 Gm13278 Gm13279 Gm13288 Gm13289 Gm13290
          Gene Name:
          interferon zeta
          predicted gene 13272
          predicted gene 13276
          predicted gene 13277
          predicted gene 13278
          predicted gene 13279
          predicted gene 13288
          predicted gene 13289
          predicted gene 13290
          Gene Aliases:
          6030405N23Rik, BGIF, ENSMUSG00000073810, Lmtn
          OTTMUSG00000011265
          OTTMUSG00000011269
          OTTMUSG00000011270
          OTTMUSG00000011272
          OTTMUSG00000011273
          OTTMUSG00000011289
          OTTMUSG00000011290
          OTTMUSG00000011291
          Chromosome Location:
          Chr.4: 88782131 - 88783592 on Build GRCm38
          Chromosome Location:
          Chr.4: 88779850 - 88780660 on Build GRCm38
          Chromosome Location:
          Chr.4: 88785054 - 88786515 on Build GRCm38
          Chromosome Location:
          Chr.4: 88787977 - 88789438 on Build GRCm38
          Chromosome Location:
          Chr.4: 88790895 - 88792356 on Build GRCm38
          Chromosome Location:
          Chr.4: 88797237 - 88798204 on Build GRCm38
          Chromosome Location:
          Chr.4: 88805531 - 88808380 on Build GRCm38
          Chromosome Location:
          Chr.4: 88776762 - 88777729 on Build GRCm38
          Chromosome Location:
          Chr.4: 88773834 - 88774801 on Build GRCm38
          UniGene ID:
          Mm.368232
          Mm.368246
          Mm.474424
          Mm.473677
          Mm.149203
          Mm.489586
          Mm.487241
          Mm.380019
          Mm.380019
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          -
          -
          -
          -
          siRNA ID s232936

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Ifnz
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_197889.2 NP_922871.1 1 1421
          GenBank mRNA AK046550.1 - 1 1422
          Gene Symbol: Gm13272
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001161608.1 NP_001155080.1 1 766
          Gene Symbol: Gm13276
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001085532.3 NP_001079001.1 1 1421
          Gene Symbol: Gm13277
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001098840.3 NP_001092310.1 1 1421
          Gene Symbol: Gm13278
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001098841.3 NP_001092311.1 1 1421
          Gene Symbol: Gm13279
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001243166.1 NP_001230095.1 1 926
          Gene Symbol: Gm13288
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001243167.1 NP_001230096.1 1 1421
          GenBank mRNA AK031317.1 - 1 1422
          Gene Symbol: Gm13289
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001243165.1 NP_001230094.1 926
          Gene Symbol: Gm13290
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001243155.1 NP_001230084.1 1 926
          GenBank mRNA AB024521.1 - 1 926

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          interferon superfamily

          Gene Ontology Categories:

          Function(s) Process(es)

          adaptive immune response
          T cell activation involved in immune response
          natural killer cell activation involved in immune response
          humoral immune response
          cytokine-mediated signaling pathway
          B cell differentiation
          positive regulation of peptidyl-serine phosphorylation of STAT protein
          B cell proliferation
          response to exogenous dsRNA
          regulation of cell cycle
          apoptotic signaling pathway
          biological_process
          cytokine activity
          type I interferon receptor binding
          molecular_function

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-znbv5:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0