Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other INS2 SIRNA Assays ›
          Gene Symbol:
          Ins2
          Gene Name:
          insulin II
          Gene Aliases:
          AA986540, Ins-2, InsII, Mody, Mody4, proinsulin
          Chromosome Location:
          Chr.7: 142678656 - 142679708 on Build GRCm38
          UniGene ID:
          Mm.4946
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID s233025

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Ins2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001185083.1 NP_001172012.1 2, 3 250
          NM_001185084.1 NP_001172013.1 1, 2 364
          NM_008387.4 NP_032413.1 2, 3 262
          GenBank mRNA AK007612.1 - 2, 3 263
          AY899305.1 - 1, 2 329
          BC099934.1 - 2, 3 235 6432976
          BC132650.1 - 2, 3 235 40130927
          BC132652.1 - 2, 3 235 40130929
          BC145554.1 - 2, 3 223 9054118
          GQ915612.1 - 2, 3 250

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_019130, BP485158, BP472188.

          Panther Classification:

          Molecular Function -

          growth factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          MAPK cascade
          activation of MAPK activity
          negative regulation of acute inflammatory response
          carbohydrate metabolic process
          glucose metabolic process
          regulation of cellular amino acid metabolic process
          acute-phase response
          ER overload response
          G-protein coupled receptor signaling pathway
          activation of JUN kinase activity
          myoblast fusion
          positive regulation of cell proliferation
          insulin receptor signaling pathway
          regulation of gene expression
          positive regulation of phosphatidylinositol 3-kinase signaling
          myotube differentiation
          glucose transport
          lipid catabolic process
          regulation of transmembrane transporter activity
          activation of protein kinase B activity
          positive regulation of cellular protein metabolic process
          negative regulation of protein oligomerization
          regulation of protein localization
          negative regulation of NAD(P)H oxidase activity
          wound healing
          negative regulation of protein catabolic process
          regulation of phosphorylation
          glucose homeostasis
          regulation of protein binding
          positive regulation of MAPK cascade
          positive regulation of phosphatidylinositol 3-kinase activity
          positive regulation of glycogen biosynthetic process
          positive regulation of DNA replication
          negative regulation of glycogen catabolic process
          positive regulation of glycolytic process
          positive regulation of mitotic nuclear division
          negative regulation of proteolysis
          negative regulation of fatty acid metabolic process
          positive regulation of glucose import
          positive regulation of insulin receptor signaling pathway
          alpha-beta T cell activation
          regulation of protein secretion
          negative regulation of protein secretion
          positive regulation of cytokine secretion
          positive regulation of peptidyl-tyrosine phosphorylation
          negative regulation of lipid catabolic process
          positive regulation of NF-kappaB transcription factor activity
          positive regulation of protein kinase B signaling
          fatty acid homeostasis
          canonical Wnt signaling pathway
          negative regulation of respiratory burst involved in inflammatory response
          positive regulation of respiratory burst
          intrinsic apoptotic signaling pathway in response to endoplasmic reticulum stress
          positive regulation of protein localization to nucleus
          negative regulation of feeding behavior
          protease binding
          insulin receptor binding
          insulin-like growth factor receptor binding
          hormone activity
          protein complex binding
          identical protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0