Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vmn2r47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105151.1 | NP_001098621.1 | 3 | 1268 |
Gene Symbol: Vmn2r30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009490.3 | NP_033516.3 | 3 | 1268 | ||
GenBank mRNA | AF011425.1 | - | 1307 |
Set Membership: |
G-protein coupled receptor |