Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ARHGEF9 SIRNA Assays ›
          Gene Symbol:
          ARHGEF9
          Gene Name:
          Cdc42 guanine nucleotide exchange factor 9
          Gene Aliases:
          COLLYBISTIN, EIEE8, HPEM-2, PEM-2, PEM2
          Chromosome Location:
          Chr.X: 63634967 - 63786025 on Build GRCh38
          UniGene ID:
          Hs.54697
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s23337

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ARHGEF9
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001173479.1 NP_001166950.1 2, 3 619
          NM_001173480.1 NP_001166951.1 2, 3 1025
          NM_015185.2 NP_056000.1 3, 4 1216
          XM_005262249.2 XP_005262306.1 3, 4 1278
          XM_005262250.3 XP_005262307.1 3, 4 1176
          XM_005262251.1 XP_005262308.1 3, 4 526
          XM_005262252.1 XP_005262309.1 4, 5 545
          XM_011530890.1 XP_011529192.1 4, 5 960
          XM_011530892.1 XP_011529194.1 3, 4 784
          XM_017029363.1 XP_016884852.1 4, 5 1305
          XM_017029364.1 XP_016884853.1 5, 6 1082
          XM_017029365.1 XP_016884854.1 3, 4 1183
          XM_017029366.1 XP_016884855.1 4, 5 964
          XM_017029367.1 XP_016884856.1 4, 5 953
          XM_017029368.1 XP_016884857.1 4, 5 1254
          XM_017029369.1 XP_016884858.1 3, 4 1278
          XM_017029370.1 XP_016884859.1 3, 4 1187
          XM_017029371.1 XP_016884860.1 3, 4 1176
          XM_017029372.1 XP_016884861.1 3, 4 526
          XM_017029373.1 XP_016884862.1 4, 5 1319
          XM_017029374.1 XP_016884863.1 2, 3 1007
          XM_017029375.1 XP_016884864.1 3, 4 1184
          XM_017029376.1 XP_016884865.1 3, 4 1184
          XM_017029377.1 XP_016884866.1 3, 4 1187
          XM_017029378.1 XP_016884867.1 3, 4 1278
          XM_017029379.1 XP_016884868.1 3, 4 526
          XM_017029380.1 XP_016884869.1 2, 3 1007
          GenBank mRNA AB007884.1 - 3, 4 1178
          AK289993.1 - 3, 4 630
          AK295033.1 - 2, 3 631
          AK295178.1 - 3, 4 526
          BC117406.1 - 3, 4 527 40125957

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          guanyl-nucleotide exchange factor

          Gene Ontology Categories:

          Function(s) Process(es)

          ion transmembrane transport
          regulation of Rho protein signal transduction
          positive regulation of apoptotic process
          positive regulation of GTPase activity
          regulation of small GTPase mediated signal transduction
          guanyl-nucleotide exchange factor activity
          Rho guanyl-nucleotide exchange factor activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0