Search
Search

Acrbp| Gene Symbol: Acrbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127340.1 | NP_001120812.1 | 5 | 981 | ||
| NM_016845.2 | NP_058541.2 | 5 | 981 | |||
| GenBank mRNA | AK076691.1 | - | 5 | 946 | ||
| AK132705.1 | - | 5 | 949 | |||
| AV260707.2 | - | 1 | 369 | |||
| BC011079.1 | - | 5 | 1219 | 3984550 | ||
| D17573.1 | - | 5 | 933 | |||
| D17574.1 | - | 5 | 985 | |||