Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acrbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127340.1 | NP_001120812.1 | 2 | 311 | ||
NM_016845.2 | NP_058541.2 | 2 | 311 | |||
GenBank mRNA | AK076691.1 | - | 2 | 276 | ||
AK132705.1 | - | 2 | 279 | |||
BC011079.1 | - | 2 | 549 | 3984550 | ||
BC061122.1 | - | 2 | 271 | 6773251 | ||
BC114591.1 | - | 2 | 280 | |||
BM938496.1 | - | 1 | 84 | |||
BY099028.1 | - | 2 | 311 | |||
D17573.1 | - | 2 | 263 | |||
D17574.1 | - | 2 | 315 |