Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xlr4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081642.1 | NP_001075111.1 | 8 | 779 | ||
GenBank mRNA | BC098242.1 | - | 8 | 759 | 1478336 |
Gene Symbol: Xlr4c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183094.3 | NP_898917.2 | 8 | 809 | ||
GenBank mRNA | AK166871.1 | - | 8 | 810 |