Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppig | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081086.1 | NP_001074555.1 | 13, 14 | 1518 | ||
GenBank mRNA | AK088007.1 | - | 13, 14 | 1328 | ||
AK134121.1 | - | 13, 14 | 1520 | |||
AK135178.1 | - | 14, 15 | 1649 | |||
AK135713.1 | - | 3, 4 | 301 | |||
BC016126.1 | - | 13, 14 | 1380 | |||
BC027420.1 | - | 13, 14 | 1512 | |||
BC038818.1 | - | 13, 14 | 1334 | |||
BC094926.1 | - | 13, 14 | 1372 | |||
BC150694.1 | - | 14, 15 | 1539 | 9087605 |