Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PLCB1 SIRNA Assays ›
          Gene Symbol:
          PLCB1
          Gene Name:
          phospholipase C beta 1
          Gene Aliases:
          EIEE12, PI-PLC, PLC-154, PLC-I, PLC-beta-1, PLC154, PLCB1A, PLCB1B
          Chromosome Location:
          Chr.20: 8132236 - 8884911 on Build GRCh38
          UniGene ID:
          Hs.431173
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s23358

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PLCB1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_015192.3 NP_056007.1 20 2476
          NM_182734.2 NP_877398.1 20 2476
          XM_011529199.2 XP_011527501.1 20 2436
          XM_011529200.2 XP_011527502.1 18 2100
          XM_011529201.2 XP_011527503.1 18 1910
          XM_011529203.2 XP_011527505.1 4 412
          XM_017027752.1 XP_016883241.1 20 2436
          XM_017027753.1 XP_016883242.1 20 2063
          GenBank mRNA AB011153.3 - 20 2476
          AJ278313.1 - 20 2092
          AJ278314.1 - 20 2092
          AK023689.1 - 8 1241
          AK127693.1 - 18 1891
          AK299195.1 - 20 2111
          AL137267.1 - 12 1311
          AY004175.1 - 20 2074
          BC069420.1 - 20 2092 7262405
          BC117231.1 - 20 2147 40125782

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          phospholipase

          Gene Ontology Categories:

          Function(s) Process(es)

          G2/M transition of mitotic cell cycle
          signal transduction
          G-protein coupled acetylcholine receptor signaling pathway
          glutamate receptor signaling pathway
          Wnt signaling pathway, calcium modulating pathway
          memory
          regulation of G-protein coupled receptor protein signaling pathway
          lipid catabolic process
          cerebral cortex development
          positive regulation of interleukin-12 production
          intracellular signal transduction
          interleukin-12-mediated signaling pathway
          interleukin-15-mediated signaling pathway
          positive regulation of embryonic development
          positive regulation of GTPase activity
          inositol phosphate metabolic process
          fat cell differentiation
          positive regulation of myoblast differentiation
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          positive regulation of JNK cascade
          phosphatidylinositol metabolic process
          insulin-like growth factor receptor signaling pathway
          positive regulation of developmental growth
          activation of meiosis involved in egg activation
          interleukin-1-mediated signaling pathway
          regulation of fertilization
          positive regulation of G1/S transition of mitotic cell cycle
          positive regulation of acrosome reaction
          negative regulation of monocyte extravasation
          positive regulation of CD24 biosynthetic process
          phosphatidylinositol phospholipase C activity
          signal transducer activity
          GTPase activator activity
          calcium ion binding
          protein binding
          calmodulin binding
          lamin binding
          phosphatidylinositol-4,5-bisphosphate binding
          enzyme binding
          protein homodimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-9rjdb:80/100.66.79.224:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline