Search
Search

PLCB1| Gene Symbol: PLCB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015192.3 | NP_056007.1 | 27 | 3440 | ||
| NM_182734.2 | NP_877398.1 | 27 | 3440 | |||
| XM_011529199.2 | XP_011527501.1 | 27 | 3400 | |||
| XM_011529200.2 | XP_011527502.1 | 25 | 3064 | |||
| XM_011529201.2 | XP_011527503.1 | 25 | 2874 | |||
| XM_011529203.2 | XP_011527505.1 | 11 | 1376 | |||
| XM_017027752.1 | XP_016883241.1 | 27 | 3400 | |||
| XM_017027753.1 | XP_016883242.1 | 27 | 3027 | |||
| GenBank mRNA | AB011153.3 | - | 27 | 3440 | ||
| AJ278313.1 | - | 27 | 3056 | |||
| AJ278314.1 | - | 27 | 3056 | |||
| AK023689.1 | - | 15 | 2205 | |||
| AK127693.1 | - | 25 | 2855 | |||
| AK299195.1 | - | 27 | 3075 | |||
| AL137267.1 | - | 19 | 2275 | |||
| AY004175.1 | - | 27 | 3038 | |||
| BC069420.1 | - | 27 | 3056 | 7262405 | ||
| BC117231.1 | - | 27 | 3111 | 40125782 | ||