Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rhox2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099316.1 | NP_001092786.1 | 2 | 163 | ||
GenBank mRNA | BC049546.1 | - | 163 |
Gene Symbol: Rhox2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029203.2 | NP_083479.1 | 2 | 185 | ||
GenBank mRNA | AK015997.1 | - | 2 | 186 | ||
BC132216.1 | - | 2 | 179 | 40130493 | ||
BC132220.1 | - | 2 | 179 | 40130497 | ||
BC145534.1 | - | 2 | 179 | |||
DQ058639.1 | - | 2 | 186 |
Gene Symbol: Rhox2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099318.1 | NP_001092788.1 | 185 |
Gene Symbol: Rhox2e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085348.1 | NP_001078817.1 | 185 |
Gene Symbol: Rhox2f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085356.1 | NP_001078825.1 | 2 | 185 |