Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other HIST1H2BE SIRNA Assays ›
          Gene Symbol:
          Hist1h2be Hist1h2bb Hist1h2bc Hist1h2bf Hist1h2bg Hist1h2bh Hist1h2bj Hist1h2bk Hist1h2bl Hist1h2bm Hist1h2bn Hist1h2bp Hist1h2bq Hist1h2br
          Gene Name:
          histone cluster 1, H2be
          histone cluster 1, H2bb
          histone cluster 1, H2bc
          histone cluster 1, H2bf
          histone cluster 1, H2bg
          histone cluster 1, H2bh
          histone cluster 1, H2bj
          histone cluster 1, H2bk
          histone cluster 1, H2bl
          histone cluster 1, H2bm
          histone cluster 1, H2bn
          histone cluster 1, H2bp
          histone cluster 1, H2bq
          histone cluster 1 H2br
          Gene Aliases:
          -
          H2b-143
          2610022J01Rik, H2bfs, R74621
          -
          -
          H2b-221
          -
          -
          H2b-291a
          H2b-291b
          -
          -
          Gm13646, OTTMUSG00000013203
          Gm11277, OTTMUSG00000000449
          Chromosome Location:
          Chr.13: 23583670 - 23621124 on Build GRCm38
          Chromosome Location:
          Chr.13: 23746734 - 23747223 on Build GRCm38
          Chromosome Location:
          Chr.13: 23684199 - 23692480 on Build GRCm38
          Chromosome Location:
          Chr.13: 23573760 - 23574190 on Build GRCm38
          Chromosome Location:
          Chr.13: 23571400 - 23571863 on Build GRCm38
          Chromosome Location:
          Chr.13: 23542923 - 23543444 on Build GRCm38
          Chromosome Location:
          Chr.13: 22043230 - 22043658 on Build GRCm38
          Chromosome Location:
          Chr.13: 22035821 - 22036320 on Build GRCm38
          Chromosome Location:
          Chr.13: 21715713 - 21716143 on Build GRCm38
          Chromosome Location:
          Chr.13: 21722044 - 21722526 on Build GRCm38
          Chromosome Location:
          Chr.13: 21754123 - 21754553 on Build GRCm38
          Chromosome Location:
          Chr.13: 21787480 - 21801561 on Build GRCm38
          Chromosome Location:
          Chr.13: 21806412 - 21810199 on Build GRCm38
          Chromosome Location:
          Chr.13: 21833743 - 21837530 on Build GRCm38
          UniGene ID:
          Mm.440391
          Mm.390567
          Mm.261676
          -
          Mm.261673
          Mm.352273
          Mm.371766
          Mm.358955
          -
          Mm.277425
          -
          Mm.264645
          Mm.371766
          Mm.471247
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          siRNA ID s233884

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Hist1h2be
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001177653.1 NP_001171124.1 2 311
          NM_001290530.1 NP_001277459.1 1 132
          NM_178194.4 NP_835501.1 2 337
          GenBank mRNA AK030546.1 - 2 339
          AK049948.1 - 2 312
          BC069889.1 - 1 125 4217814
          Gene Symbol: Hist1h2bb
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA BC116853.1 - 1 179 40126172
          BC116857.1 - 1 179 40126176
          Gene Symbol: Hist1h2bc
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001290380.1 NP_001277309.1 1 151
          NM_023422.3 NP_075911.2 1 151
          GenBank mRNA AK005191.1 - 1 147
          AK005407.1 - 1 146
          AK011516.1 - 1 153
          BC019673.1 - 1 134 3993954
          BC082774.1 - 1 134
          Gene Symbol: Hist1h2bf
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_178195.2 NP_835502.1 1 118
          Gene Symbol: Hist1h2bg
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_178196.4 NP_835503.1 1 151
          GenBank mRNA BC060304.1 - 1 178 4217587
          Gene Symbol: Hist1h2bh
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_178197.2 NP_835504.1 1 210
          GenBank mRNA AK158416.1 - 159
          BC092138.1 - 125 30613720
          BC132584.1 - 1 210 40130861
          BC132586.1 - 1 210 40130863
          Gene Symbol: Hist1h2bj
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA BC115792.1 - 110
          Gene Symbol: Hist1h2bk
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_175665.2 NP_783596.1 1 184
          GenBank mRNA BC147143.1 - 1 184 9056184
          BC147144.1 - 1 184 9056197
          Gene Symbol: Hist1h2bl
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_178199.2 NP_835506.1 1 118
          Gene Symbol: Hist1h2bm
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_178200.2 NP_835507.1 1 172
          GenBank mRNA BC139381.1 - 1 172 8862403
          BC139382.1 - 1 172 8862404
          Gene Symbol: Hist1h2bn
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_178201.2 NP_835508.1 1 118
          Gene Symbol: Hist1h2bp
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001290466.1 NP_001277395.1 1 145
          NM_178202.2 NP_835509.2 1 145
          GenBank mRNA AK076671.1 - 1 146
          BC061044.1 - 1 145 6742370
          M25487.1 - 1 106
          Gene Symbol: Hist1h2bq
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001097979.2 NP_001091448.2 1 146
          Gene Symbol: Hist1h2br
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001110555.1 NP_001104025.1 1 146
          GenBank mRNA AK036389.1 - 1 147
          BC011440.1 - 1 131 3989862

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001109400, NM_022647, NM_001106114, NM_001108414, XM_225384, and others.

          Panther Classification:

          Molecular Function -

          histone

          Gene Ontology Categories:

          Function(s) Process(es)

          innate immune response in mucosa
          nucleosome assembly
          antibacterial humoral response
          defense response to Gram-positive bacterium
          biological_process
          molecular_function
          DNA binding
          protein heterodimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0