Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other HIST1H2BG SIRNA Assays ›
          Gene Symbol:
          Hist1h2bg Hist1h2bb Hist1h2bc Hist1h2be Hist1h2bf Hist1h2bh Hist1h2bj Hist1h2bk Hist1h2bl Hist1h2bm Hist1h2bn Hist1h2bp Hist1h2bq Hist1h2br
          Gene Name:
          histone cluster 1, H2bg
          histone cluster 1, H2bb
          histone cluster 1, H2bc
          histone cluster 1, H2be
          histone cluster 1, H2bf
          histone cluster 1, H2bh
          histone cluster 1, H2bj
          histone cluster 1, H2bk
          histone cluster 1, H2bl
          histone cluster 1, H2bm
          histone cluster 1, H2bn
          histone cluster 1, H2bp
          histone cluster 1, H2bq
          histone cluster 1 H2br
          Gene Aliases:
          -
          H2b-143
          2610022J01Rik, H2bfs, R74621
          -
          -
          H2b-221
          -
          -
          H2b-291a
          H2b-291b
          -
          -
          Gm13646, OTTMUSG00000013203
          Gm11277, OTTMUSG00000000449
          Chromosome Location:
          Chr.13: 23571400 - 23571863 on Build GRCm38
          Chromosome Location:
          Chr.13: 23746734 - 23747223 on Build GRCm38
          Chromosome Location:
          Chr.13: 23684199 - 23692480 on Build GRCm38
          Chromosome Location:
          Chr.13: 23583670 - 23621124 on Build GRCm38
          Chromosome Location:
          Chr.13: 23573760 - 23574190 on Build GRCm38
          Chromosome Location:
          Chr.13: 23542923 - 23543444 on Build GRCm38
          Chromosome Location:
          Chr.13: 22043230 - 22043658 on Build GRCm38
          Chromosome Location:
          Chr.13: 22035821 - 22036320 on Build GRCm38
          Chromosome Location:
          Chr.13: 21715713 - 21716143 on Build GRCm38
          Chromosome Location:
          Chr.13: 21722044 - 21722526 on Build GRCm38
          Chromosome Location:
          Chr.13: 21754123 - 21754553 on Build GRCm38
          Chromosome Location:
          Chr.13: 21787480 - 21801561 on Build GRCm38
          Chromosome Location:
          Chr.13: 21806412 - 21810199 on Build GRCm38
          Chromosome Location:
          Chr.13: 21833743 - 21837530 on Build GRCm38
          UniGene ID:
          Mm.261673
          Mm.390567
          Mm.261676
          Mm.440391
          -
          Mm.352273
          Mm.371766
          Mm.358955
          -
          Mm.277425
          -
          Mm.264645
          Mm.371766
          Mm.471247
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          siRNA ID s233886

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Hist1h2bg
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_178196.4 NP_835503.1 1 150
          GenBank mRNA BC060304.1 - 1 177 4217587
          Gene Symbol: Hist1h2bb
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA BC116853.1 - 1 178 40126172
          BC116857.1 - 1 178 40126176
          Gene Symbol: Hist1h2bc
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001290380.1 NP_001277309.1 1 150
          NM_023422.3 NP_075911.2 1 150
          GenBank mRNA AK005191.1 - 1 146
          AK005407.1 - 1 145
          AK011516.1 - 1 152
          BC019673.1 - 1 133 3993954
          BC082774.1 - 1 133
          Gene Symbol: Hist1h2be
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001177653.1 NP_001171124.1 2 310
          NM_001290530.1 NP_001277459.1 1 131
          NM_178194.4 NP_835501.1 2 336
          GenBank mRNA AK030546.1 - 2 338
          AK049948.1 - 2 311
          BC069889.1 - 1 124 4217814
          Gene Symbol: Hist1h2bf
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_178195.2 NP_835502.1 1 117
          Gene Symbol: Hist1h2bh
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_178197.2 NP_835504.1 1 209
          GenBank mRNA AK158416.1 - 158
          BC092138.1 - 124 30613720
          BC132584.1 - 1 209 40130861
          BC132586.1 - 1 209 40130863
          Gene Symbol: Hist1h2bj
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA BC115792.1 - 109
          Gene Symbol: Hist1h2bk
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_175665.2 NP_783596.1 1 183
          GenBank mRNA BC147143.1 - 1 183 9056184
          BC147144.1 - 1 183 9056197
          Gene Symbol: Hist1h2bl
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_178199.2 NP_835506.1 1 117
          Gene Symbol: Hist1h2bm
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_178200.2 NP_835507.1 1 171
          GenBank mRNA BC139381.1 - 1 171 8862403
          BC139382.1 - 1 171 8862404
          Gene Symbol: Hist1h2bn
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_178201.2 NP_835508.1 1 117
          Gene Symbol: Hist1h2bp
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001290466.1 NP_001277395.1 1 144
          NM_178202.2 NP_835509.2 1 144
          GenBank mRNA AK076671.1 - 1 145
          BC061044.1 - 1 144 6742370
          M25487.1 - 1 105
          Gene Symbol: Hist1h2bq
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001097979.2 NP_001091448.2 1 145
          Gene Symbol: Hist1h2br
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001110555.1 NP_001104025.1 1 145
          GenBank mRNA AK036389.1 - 1 146
          BC011440.1 - 1 130 3989862

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001109400, NM_022647, NM_001106114, NM_001108414, XM_225384, and others.

          Panther Classification:

          Molecular Function -

          histone

          Gene Ontology Categories:

          Function(s) Process(es)

          innate immune response in mucosa
          nucleosome assembly
          antibacterial humoral response
          defense response to Gram-positive bacterium
          biological_process
          molecular_function
          DNA binding
          protein heterodimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-njpwx:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0