Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnfsf13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159505.1 | NP_001152977.1 | 1 | 413 | ||
NM_023517.2 | NP_076006.2 | 1 | 413 | |||
GenBank mRNA | AK009514.1 | - | 1 | 415 | ||
AK040667.1 | - | 1 | 191 | |||
AK134028.1 | - | 1 | 119 | |||
AK170351.1 | - | 1 | 420 | |||
BC069900.1 | - | 1 | 372 | 5290965 |