Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KAZN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017999.2 | NP_001017999.1 | 8 | 1144 | ||
NM_001018000.3 | NP_001018000.1 | 8 | 1333 | |||
NM_001018001.2 | NP_001018001.1 | 8 | 1134 | |||
NM_015209.2 | NP_056024.1 | 8 | 1396 | |||
NM_201628.2 | NP_963922.2 | 8 | 1396 | |||
XM_005245795.4 | XP_005245852.1 | 10 | 1651 | |||
XM_011541074.2 | XP_011539376.1 | 10 | 1658 | |||
XM_011541075.2 | XP_011539377.1 | 8 | 1314 | |||
XM_011541076.2 | XP_011539378.1 | 8 | 1459 | |||
XM_011541077.2 | XP_011539379.1 | 8 | 2438 | |||
XM_011541080.2 | XP_011539382.1 | 10 | 1651 | |||
XM_011541081.2 | XP_011539383.1 | 2 | 138 | |||
XM_017000770.1 | XP_016856259.1 | 10 | 1646 | |||
XM_017000771.1 | XP_016856260.1 | 9 | 1319 | |||
XM_017000772.1 | XP_016856261.1 | 8 | 998 | |||
GenBank mRNA | AB015329.1 | - | 7 | 786 | ||
AK293806.1 | - | 3 | 202 | |||
AY505119.1 | - | 9 | 1324 | |||
AY505120.1 | - | 8 | 1333 | |||
AY505121.1 | - | 8 | 1134 | |||
AY505122.1 | - | 8 | 1144 | |||
BC035501.2 | - | 8 | 1332 | 4556194 | ||
DR003677.1 | - | 3 | 119 |