siRNA Details
Gene Symbol: Snrpn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001082961.1 | NP_001076430.1 | 5 | 494 | ||
NM_001082962.1 | NP_001076431.1 | 5 | 538 | |||
NM_013670.3 | NP_038698.1 | 4 | 432 | |||
GenBank mRNA | AK010671.1 | - | 4 | 469 | ||
AK013607.1 | - | 4 | 389 | |||
AK144386.1 | - | 4 | 388 | |||
BC019589.1 | - | 4 | 380 | 3484323 | ||
BC024880.1 | - | 4 | 371 | 3987218 | ||
BC069894.1 | - | 4 | 369 | |||
BU614958.1 | - | 5 | 538 | |||
S40304.1 | - | 1 | 77 | |||
X62648.1 | - | 1 | 77 | |||
X63730.1 | - | 1 | 30 |
Gene Symbol: Snurf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033174.3 | NP_149409.1 | 4 | 432 | ||
GenBank mRNA | AF101042.1 | - | 4 | 399 | ||
BC132082.1 | - | 4 | 382 | 40130359 | ||
BC132084.1 | - | 4 | 382 | 40130361 |

More Information
Panther Classification:
Molecular Function -




