Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other LOC101055863 SIRNA Assays ›
          Gene Symbol:
          LOC101055863 Gm11236 Gm11237 Gm11487 Gm11757 Gm11758 Gm13871 Gm428 LOC100502896 LOC100862015
          Gene Name:
          uncharacterized LOC101055863
          predicted gene 11236
          predicted gene 11237
          predicted gene 11487
          predicted gene 11757
          predicted gene 11758
          predicted gene 13871
          predicted gene 428
          uncharacterized LOC100502896
          uncharacterized LOC100862015
          Gene Aliases:
          -
          OTTMUSG00000000280
          OTTMUSG00000000281
          OTTMUSG00000001246
          OTTMUSG00000004012
          OTTMUSG00000004013
          OTTMUSG00000014862
          -
          -
          -
          Chromosome Location:
          Chr.4: 73666018 - 73672398 on Build GRCm38
          Chromosome Location:
          Chr.4: 73620648 - 73627029 on Build GRCm38
          Chromosome Location:
          Chr.4: 73605544 - 73611922 on Build GRCm38
          Chromosome Location:
          Chr.4: 73401032 - 73407404 on Build GRCm38
          Chromosome Location:
          Chr.4: 73887066 - 73891080 on Build GRCm38
          Chromosome Location:
          Chr.4: 73872219 - 73876266 on Build GRCm38
          Chromosome Location:
          Chr.4: 73901906 - 73905921 on Build GRCm38
          Chromosome Location:
          Chr.4: 73681151 - 73687559 on Build GRCm38
          Chromosome Location:
          Chr.4: 73650910 - 73657292 on Build GRCm38
          Chromosome Location:
          Chr.4: 73635785 - 73642163 on Build GRCm38
          UniGene ID:
          Mm.331671
          -
          Mm.331671
          Mm.348294
          Mm.389560
          Mm.457901
          Mm.457901
          Mm.331671
          Mm.389560
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          siRNA ID s234221

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: LOC101055863
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001277487.1 NP_001264416.1 223
          GenBank mRNA CN719955.1 - 201
          Gene Symbol: Gm11236
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001277575.1 NP_001264504.1 223
          Gene Symbol: Gm11237
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001256481.2 NP_001243410.2 3 223
          GenBank mRNA BY725321.1 - 307
          Gene Symbol: Gm11487
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001013393.1 NP_001013411.1 3 307
          Gene Symbol: Gm11757
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001085538.2 NP_001079007.1 277
          GenBank mRNA BG083055.2 - 130
          Gene Symbol: Gm11758
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001097978.2 NP_001091447.2 309
          GenBank mRNA BC172045.1 - 3 277 9054331
          Gene Symbol: Gm13871
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001177578.2 NP_001171049.1 3 277
          Gene Symbol: Gm428
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001081644.1 NP_001075113.1 309
          GenBank mRNA AK139501.1 - 311
          Gene Symbol: LOC100502896
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001277512.1 NP_001264441.1 223
          GenBank mRNA DV648387.1 - 222
          Gene Symbol: LOC100862015
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001277531.1 NP_001264460.1 223

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-st8h5:80/100.66.79.144:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0