Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other GAPDH SIRNA Assays ›
Gene Symbol:
Gapdh
Gm5069
Gene Name:
glyceraldehyde-3-phosphate dehydrogenase
glyceraldehyde-3-phosphate dehydrogenase pseudogene
Gene Aliases:
Gapd
EG277333
Chromosome Location:
Chr.6: 125161338 - 125166511 on Build GRCm38
Chromosome Location:
Chr.1: 180326969 - 180330549 on Build GRCm38
UniGene ID:
Mm.304088
Mm.389223
Species:
Mouse
Species Specific ID (Flybase ID):
-
-
siRNA ID
s234321
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
5,354.00
Your Price
Online offer:
5354.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Gapdh
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_008084.3
NP_032110.1
1, 2
231
GenBank mRNA
AK002273.1
-
1, 2
64
AK081405.1
-
1, 2
64
AK140794.1
-
1, 2
64
AK146435.1
-
1, 2
64
AK147738.1
-
1, 2
64
AK147891.1
-
1, 2
64
AK160399.1
-
1, 2
64
AK160753.1
-
1, 2
61
AK164415.1
-
1, 2
62
AK184442.1
-
1, 2
32
AK189223.1
-
1, 2
32
AK191938.1
-
1, 2
32
AK192040.1
-
1, 2
32
AK198590.1
-
1, 2
32
AK199549.1
-
1, 2
32
AK199608.1
-
1, 2
32
AK201453.1
-
32
AK201736.1
-
1, 2
32
AY618199.1
-
562
BC020407.1
-
1, 2
52
BC023196.1
-
1, 2
62
BC082592.1
-
1, 2
41
30634381
BC083065.1
-
1, 2
40
5371133
BC083079.1
-
1, 2
46
6493407
BC083080.1
-
1, 2
48
6494145
BC083149.1
-
27
30459585
BC085274.1
-
49
6530979
BC085275.1
-
1, 2
58
30444086
BC085315.1
-
1, 2
43
30440429
BC091768.1
-
27
4159824
BC092252.1
-
1, 2
50
5044783
BC092264.1
-
52
2655526
BC092267.1
-
38
4010884
BC092294.1
-
41
5068049
BC093508.1
-
21
4975394
BC094037.1
-
28
5009636
BC095932.1
-
1, 2
52
30733532
BC096440.1
-
1, 2
52
2649847
BC096590.1
-
52
6486174
BC110311.1
-
1, 2
53
5340298
BC145810.1
-
1, 2
34
40131113
BC145812.1
-
1, 2
34
40131115
BU504528.1
-
49
CJ155387.1
-
1, 2
231
GU214026.1
-
1, 2
62
M32599.1
-
1, 2
37
Gene Symbol: Gm5069
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
GenBank mRNA
BC064681.1
-
2
104
Back To Top
More Information
Panther Classification:
Molecular Function -
dehydrogenase
Gene Ontology Categories:
Function(s)
Process(es)
microtubule cytoskeleton organization
carbohydrate metabolic process
gluconeogenesis
glycolytic process
apoptotic process
multicellular organism development
negative regulation of translation
peptidyl-cysteine S-trans-nitrosylation
protein stabilization
neuron apoptotic process
oxidation-reduction process
response to ammonium ion
cellular response to interferon-gamma
glyceraldehyde-3-phosphate dehydrogenase (NAD+) (phosphorylating) activity
microtubule binding
peptidyl-cysteine S-nitrosylase activity
identical protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0