Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 企業服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other GM20736 SIRNA Assays ›
          Gene Symbol:
          Gm20736 1700040F15Rik D630029K05Rik Gm20857 Gm20871 Pmel Sly
          Gene Name:
          predicted gene, 20736
          RIKEN cDNA 1700040F15 gene
          RIKEN cDNA D630029K05 gene
          Sycp3 like Y-linked pseudogene
          Sycp3 like Y-linked pseudogene
          premelanosome protein
          Sycp3 like Y-linked
          Gene Aliases:
          -
          -
          AI314111, AI875589
          -
          -
          D10H12S53E, D12S53Eh, Pmel17, Si, Silv, gp100, gp87
          -
          Chromosome Location:
          Chr.Y: 65377690 - 65403913 on Build GRCm38
          Chromosome Location:
          Chr.Y: 60241398 - 60297384 on Build GRCm38
          Chromosome Location:
          Chr.10: 116964740 - 116969108 on Build GRCm38
          Chromosome Location:
          Chr.Y: 25214873 - 25240906 on Build GRCm38
          Chromosome Location:
          Chr.Y: 59606710 - 59632758 on Build GRCm38
          Chromosome Location:
          Chr.10: 128705168 - 128720238 on Build GRCm38
          Chromosome Location:
          Chr.Y: 57187206 - 57213358 on Build GRCm38
          UniGene ID:
          Mm.391354
          Mm.293024
          Mm.27138
          Mm.359525
          Mm.426191
          Mm.16756
          Mm.304354
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          -
          -
          siRNA ID s234347

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Gm20736
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001037748.1 NP_001032837.2 6, 7 519
          GenBank mRNA BC100464.1 - 6, 7 519 6705131
          Gene Symbol: 1700040F15Rik
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AK076905.1 - 490
          AK077064.1 - 9, 10 3152
          FJ541082.1 - 567
          FJ541086.1 - 566
          FJ541087.1 - 597
          FJ541088.1 - 380
          FJ541094.1 - 313
          FJ541095.1 - 400
          FJ541098.1 - 364
          FJ541100.1 - 299
          FJ541102.1 - 180
          Gene Symbol: D630029K05Rik
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA FJ541076.1 - 630
          Gene Symbol: Gm20857
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA BU945722.1 - 556
          Gene Symbol: Gm20871
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA BC099537.1 - 493 30916067
          Gene Symbol: Pmel
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA FJ541090.1 - 247
          Gene Symbol: Sly
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_201530.2 NP_963288.2 8, 9 606
          GenBank mRNA BC049626.1 - 8, 9 606 6705850

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          spermatid development
          single fertilization
          sex determination
          regulation of gene expression
          spermatid differentiation
          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-7l2qm:80/100.66.75.127:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline