Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ADGRL2 SIRNA Assays ›
Gene Symbol:
ADGRL2
Gene Name:
adhesion G protein-coupled receptor L2
Gene Aliases:
CIRL2, CL2, LEC1, LPHH1, LPHN2
Chromosome Location:
Chr.1: 81306155 - 81992738 on Build GRCh38
UniGene ID:
Hs.24212
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s23435
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ADGRL2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001297704.1
NP_001284633.1
6
714
NM_001297705.1
NP_001284634.1
5
749
NM_001297706.1
NP_001284635.1
5
749
NM_012302.3
NP_036434.1
5
749
XM_005270666.4
XP_005270723.1
5
1470
XM_005270668.2
XP_005270725.1
5
766
XM_006710485.3
XP_006710548.1
6
1486
XM_006710488.2
XP_006710551.1
6
780
XM_006710489.3
XP_006710552.1
6
671
XM_017000782.1
XP_016856271.1
10
1354
XM_017000783.1
XP_016856272.1
7
765
XM_017000784.1
XP_016856273.1
6
670
XM_017000785.1
XP_016856274.1
6
777
XM_017000786.1
XP_016856275.1
6
781
XM_017000787.1
XP_016856276.1
6
692
XM_017000788.1
XP_016856277.1
7
707
XM_017000789.1
XP_016856278.1
6
691
XM_017000790.1
XP_016856279.1
7
706
XM_017000791.1
XP_016856280.1
5
770
XM_017000792.1
XP_016856281.1
6
705
XM_017000793.1
XP_016856282.1
7
708
XM_017000794.1
XP_016856283.1
7
707
XM_017000795.1
XP_016856284.1
7
706
XM_017000796.1
XP_016856285.1
5
730
XM_017000797.1
XP_016856286.1
5
675
XM_017000798.1
XP_016856287.1
5
737
GenBank mRNA
AF104266.1
-
5
749
AF104938.1
-
5
749
AF104939.1
-
5
749
AJ131581.1
-
6
713
Back To Top
More Information
Set Membership:
G-protein coupled receptor
Panther Classification:
Molecular Function -
G-protein coupled receptor
Gene Ontology Categories:
Function(s)
Process(es)
cell surface receptor signaling pathway
G-protein coupled receptor signaling pathway
G-protein coupled receptor activity
latrotoxin receptor activity
carbohydrate binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5c7f9f6c68-kz7s5:80/10.239.165.170:80.
git-commit: b16971a8df2afcd8df684aafbc9bf1e9f45e42d4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.6.0-2024.04.15-1.0