Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RHOX4F SIRNA Assays ›
          Gene Symbol:
          Rhox4f Rhox4a Rhox4b Rhox4c Rhox4d Rhox4e Rhox4g
          Gene Name:
          reproductive homeobox 4F
          reproductive homeobox 4A
          reproductive homeobox 4B
          reproductive homeobox 4C
          reproductive homeobox 4D
          reproductive homeobox 4E
          reproductive homeobox 4G
          Gene Aliases:
          Rhox4.6, Rhox4e, Rhox4h
          Rhox4, Rhox4.1
          5430432L21Rik, Ehox, Rhox4.2, Rhox4a
          Rhox4.3, Rhox4b
          Rhox4.4, Rhox4c
          Rhox4, Rhox4.5, Rhox4d
          Rhox4.6, Rhox4.7, Rhox4f
          Chromosome Location:
          Chr.X: 37602895 - 37607686 on Build GRCm38
          Chromosome Location:
          Chr.X: 37265318 - 37270136 on Build GRCm38
          Chromosome Location:
          Chr.X: 37432494 - 37437278 on Build GRCm38
          Chromosome Location:
          Chr.X: 37480337 - 37485124 on Build GRCm38
          Chromosome Location:
          Chr.X: 37514389 - 37519176 on Build GRCm38
          Chromosome Location:
          Chr.X: 37557412 - 37562282 on Build GRCm38
          Chromosome Location:
          Chr.X: 37646501 - 37651327 on Build GRCm38
          UniGene ID:
          Mm.491809
          Mm.296330
          Mm.327419
          Mm.296444
          Mm.381921
          Mm.379498
          Mm.491809
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          -
          -
          siRNA ID s234363

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Rhox4f
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001039696.1 NP_001034785.1 4 783
          GenBank mRNA AJ972670.1 - 4 783
          Gene Symbol: Rhox4a
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001039688.3 NP_001034777.1 4 745
          GenBank mRNA AJ972665.1 - 4 745
          AK017380.1 - 4 660
          BC057925.1 - 4 660 5292192
          BC116370.1 - 4 693 40100097
          BC116371.1 - 4 692 40100098
          BC145382.1 - 4 646 9053940
          BC171959.1 - 4 646 9054245
          DQ058641.1 - 4 720
          Gene Symbol: Rhox4b
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_021300.2 NP_067275.1 4 740
          GenBank mRNA AF265350.1 - 4 728
          AJ972666.1 - 4 740
          BC172126.1 - 4 687 9054796
          Gene Symbol: Rhox4c
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001039689.1 NP_001034778.1 4 757
          GenBank mRNA AJ972667.1 - 4 757
          Gene Symbol: Rhox4d
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001039695.1 NP_001034784.1 613
          GenBank mRNA AJ972668.1 - 613
          BC147696.1 - 4 687 9056669
          Gene Symbol: Rhox4e
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_201236.3 NP_957688.2 4 831
          GenBank mRNA AJ972669.1 - 4 831
          BC094891.1 - 4 793 30019143
          BC172128.1 - 687 9054798
          Gene Symbol: Rhox4g
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001039698.1 NP_001034787.1 4 832
          GenBank mRNA AJ972671.1 - 4 832

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s)

          biological_process

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0