Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RHOX4C SIRNA Assays ›
          Gene Symbol:
          Rhox4c Rhox4a Rhox4b Rhox4d Rhox4e Rhox4f Rhox4g
          Gene Name:
          reproductive homeobox 4C
          reproductive homeobox 4A
          reproductive homeobox 4B
          reproductive homeobox 4D
          reproductive homeobox 4E
          reproductive homeobox 4F
          reproductive homeobox 4G
          Gene Aliases:
          Rhox4.3, Rhox4b
          Rhox4, Rhox4.1
          5430432L21Rik, Ehox, Rhox4.2, Rhox4a
          Rhox4.4, Rhox4c
          Rhox4, Rhox4.5, Rhox4d
          Rhox4.6, Rhox4e, Rhox4h
          Rhox4.6, Rhox4.7, Rhox4f
          Chromosome Location:
          Chr.X: 37480337 - 37485124 on Build GRCm38
          Chromosome Location:
          Chr.X: 37265318 - 37270136 on Build GRCm38
          Chromosome Location:
          Chr.X: 37432494 - 37437278 on Build GRCm38
          Chromosome Location:
          Chr.X: 37514389 - 37519176 on Build GRCm38
          Chromosome Location:
          Chr.X: 37557412 - 37562282 on Build GRCm38
          Chromosome Location:
          Chr.X: 37602895 - 37607686 on Build GRCm38
          Chromosome Location:
          Chr.X: 37646501 - 37651327 on Build GRCm38
          UniGene ID:
          Mm.296444
          Mm.296330
          Mm.327419
          Mm.381921
          Mm.379498
          Mm.491809
          Mm.491809
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          -
          -
          siRNA ID s234383

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Rhox4c
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001039689.1 NP_001034778.1 4 757
          GenBank mRNA AJ972667.1 - 4 757
          Gene Symbol: Rhox4a
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001039688.3 NP_001034777.1 4 745
          GenBank mRNA AJ972665.1 - 4 745
          AK017380.1 - 4 660
          BC057925.1 - 4 660 5292192
          BC116370.1 - 4 693 40100097
          BC116371.1 - 4 692 40100098
          BC145382.1 - 4 646 9053940
          BC171959.1 - 4 646 9054245
          DQ058641.1 - 4 720
          Gene Symbol: Rhox4b
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_021300.2 NP_067275.1 4 740
          GenBank mRNA AF265350.1 - 4 728
          AJ972666.1 - 4 740
          BC172126.1 - 4 687 9054796
          Gene Symbol: Rhox4d
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001039695.1 NP_001034784.1 613
          GenBank mRNA AJ972668.1 - 613
          BC147696.1 - 4 687 9056669
          Gene Symbol: Rhox4e
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_201236.3 NP_957688.2 4 831
          GenBank mRNA AJ972669.1 - 4 831
          BC094891.1 - 4 793 30019143
          BC172128.1 - 687 9054798
          Gene Symbol: Rhox4f
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001039696.1 NP_001034785.1 4 783
          GenBank mRNA AJ972670.1 - 4 783
          Gene Symbol: Rhox4g
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001039698.1 NP_001034787.1 4 832
          GenBank mRNA AJ972671.1 - 4 832

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s)

          biological_process

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0