Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other UBE2D3 SIRNA Assays ›
          Gene Symbol:
          Ube2d3 Ube2d2a
          Gene Name:
          ubiquitin-conjugating enzyme E2D 3
          ubiquitin-conjugating enzyme E2D 2A
          Gene Aliases:
          1100001F19Rik, 9430029A22Rik, AA414951
          1500034D03Rik, Ubc2e, Ube2d2, ubc4
          Chromosome Location:
          Chr.3: 135438244 - 135467180 on Build GRCm38
          Chromosome Location:
          Chr.18: 35771559 - 35807172 on Build GRCm38
          UniGene ID:
          Mm.391601
          Mm.180052
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID s234443

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Ube2d3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_025356.4 NP_079632.1 6 493
          GenBank mRNA AK003163.1 - 6 444
          AK004001.1 - 6 503
          AK009276.1 - 6 435
          AK009936.1 - 6 432
          AK032885.1 - 6 503
          AK049906.1 - 6 494
          AK088448.1 - 6 463
          AK151295.1 - 6 607
          AK151594.1 - 6 514
          AK152231.1 - 6 514
          AK152541.1 - 6 487
          AK153100.1 - 6 607
          AK153111.1 - 6 515
          AK153410.1 - 6 514
          AK153423.1 - 6 553
          AK159125.1 - 6 553
          AK159546.1 - 6 489
          AK159963.1 - 6 487
          AK165235.1 - 5 734
          AK167077.1 - 6 503
          AK167138.1 - 6 456
          AK167650.1 - 6 491
          AK167801.1 - 6 515
          AK169177.1 - 5 865
          BC023266.1 - 5 258
          BC024351.1 - 7 542
          BC057941.1 - 6 474 5323636
          BC098220.1 - 6 508 963757
          Gene Symbol: Ube2d2a
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_019912.2 NP_064296.1 5 686
          GenBank mRNA AK005355.1 - 6 638
          AK136197.1 - 5 624
          AK139623.1 - 5 652
          AK146659.1 - 3 818
          AK147568.1 - 4 6372
          AK148089.1 - 5 508
          AK160567.1 - 5 659
          AK162093.1 - 5 491
          AK172088.1 - 5 513
          AK216283.1 - 1 66
          BC003923.1 - 5 667 3485259
          U62483.1 - 5 642

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Human: NM_003339, NM_181838, BQ186099, BC033349, AK001428, and others.
          This siRNA also targets the following transcripts in Rat: NM_031001, NM_031237, NM_001037292, XM_006233334, XM_006233336, and others.

          Panther Classification:

          Molecular Function -

          ubiquitin-protein ligase

          Gene Ontology Categories:

          Function(s) Process(es)

          protein polyubiquitination
          DNA repair
          ubiquitin-dependent protein catabolic process
          protein monoubiquitination
          apoptotic process
          cellular response to DNA damage stimulus
          protein ubiquitination
          proteasome-mediated ubiquitin-dependent protein catabolic process
          protein autoubiquitination
          protein K48-linked ubiquitination
          protein K11-linked ubiquitination
          positive regulation of protein targeting to mitochondrion
          nucleotide binding
          ubiquitin-protein transferase activity
          protein binding
          ATP binding
          ligase activity
          ubiquitin conjugating enzyme activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-2rf75:80/100.66.79.83:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0